Assay Details
| Gene Symbol: Cbfa2t3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109873.1 | NP_001103343.1 | 5 - 6 | 876 | 78 | |
| NM_009824.2 | NP_033954.2 | 5 - 6 | 857 | 78 | ||
| NM_177289.1 | NP_796263.1 | 4 - 5 | 798 | 78 | ||
| GenBank mRNA | AF038029.1 | - | 5 - 6 | 739 | 78 | |
| AK033680.1 | - | 4 - 5 | 695 | 78 | ||
| AK041887.1 | - | 5 - 6 | 878 | 78 | ||
| BC141396.1 | - | 5 - 6 | 686 | 9056009 | 78 | |
| BC151156.1 | - | 5 - 6 | 9088044 | 78 | ||
More Information
Set Membership: |
|

Cbfa2t3
Genomic Map