Assay Details
| Gene Symbol: Ctla4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009843.4 | NP_033973.2 | 2 - 3 | 609 | 68 | |
| GenBank mRNA | AK138891.1 | - | 2 - 3 | 542 | 68 | |
| BC042741.1 | - | 2 - 3 | 609 | 4936052 | 68 | |
| X05719.1 | - | 2 - 3 | 520 | 68 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ctla4
Genomic Map