Assay Details
| Gene Symbol: Cdc25c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK141737.1 | - | 8 - 9 | 907 | 60 | |
| AK168287.1 | - | 8 - 9 | 902 | 60 | ||
| BC004669.1 | - | 7 - 8 | 611 | 3498846 | 60 | |
| L16926.1 | - | 7 - 8 | 1089 | 60 | ||
| U15562.1 | - | 8 - 9 | 873 | 60 | ||
More Information
Set Membership: |
|

Cdc25c
Genomic Map