Assay Details
| Gene Symbol: Cdh1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009864.2 | NP_033994.1 | 11 - 12 | 1845 | 106 | |
| GenBank mRNA | AK076369.1 | - | 11 - 12 | 1847 | 106 | |
| BC098501.1 | - | 11 - 12 | 1834 | 30023851 | 106 | |
| X06115.1 | - | 11 - 12 | 1786 | 106 | ||
| X06339.1 | - | 8 - 9 | 1199 | 106 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cdh1
Genomic Map