Assay Details
| Gene Symbol: Cdh1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009864.2 | NP_033994.1 | 13 - 14 | 2298 | 87 | |
| GenBank mRNA | AK076369.1 | - | 13 - 14 | 2300 | 87 | |
| AK166267.1 | - | 13 - 14 | 2300 | 87 | ||
| BC098501.1 | - | 13 - 14 | 2287 | 30023851 | 87 | |
| X06115.1 | - | 13 - 14 | 2239 | 87 | ||
| X06339.1 | - | 10 - 11 | 1652 | 87 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cdh1
Genomic Map