Assay Details
| Gene Symbol: Cebpz | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024806.2 | NP_001019977.1 | 8 - 9 | 2420 | 83 | |
| GenBank mRNA | AK136173.1 | - | 8 - 9 | 2401 | 83 | |
| AK155990.1 | - | 8 - 9 | 2403 | 83 | ||
| AK158771.1 | - | 8 - 9 | 2397 | 83 | ||
| AK159081.1 | - | 8 - 9 | 2417 | 83 | ||
| U19891.1 | - | 8 - 9 | 2391 | 83 | ||
| U19892.1 | - | 8 - 9 | 2105 | 83 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cebpz
Genomic Map