Assay Details
| Gene Symbol: Cnn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009922.4 | NP_034052.3 | 2 - 3 | 286 | 65 | |
| GenBank mRNA | Z19542.1 | - | 2 - 3 | 280 | 65 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: Cnn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009922.4 | NP_034052.3 | 2 - 3 | 286 | 65 | |
| GenBank mRNA | Z19542.1 | - | 2 - 3 | 280 | 65 | |
Set Membership: |
|