Assay Details
| Gene Symbol: Cnn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009922.4 | NP_034052.3 | 3 - 4 | 359 | 129 | |
| GenBank mRNA | AK035548.1 | - | 3 - 4 | 332 | 129 | |
| BC138863.1 | - | 3 - 4 | 291 | 8861885 | 129 | |
| BC138864.1 | - | 3 - 4 | 291 | 8861886 | 129 | |
| BC145446.1 | - | 3 - 4 | 291 | 129 | ||
| Z19542.1 | - | 3 - 4 | 353 | 129 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cnn1
Genomic Map