Assay Details
| Gene Symbol: Cnn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009922.4 | NP_034052.3 | 4 - 5 | 493 | 78 | |
| GenBank mRNA | AK035548.1 | - | 4 - 5 | 466 | 78 | |
| BC138863.1 | - | 4 - 5 | 425 | 8861885 | 78 | |
| BC138864.1 | - | 4 - 5 | 425 | 8861886 | 78 | |
| BC145446.1 | - | 4 - 5 | 425 | 78 | ||
| Z19542.1 | - | 4 - 5 | 487 | 78 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cnn1
Genomic Map