Assay Details
| Gene Symbol: Csnk2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK002903.1 | - | 5 - 6 | 492 | 60 | |
| AK010730.1 | - | 5 - 6 | 489 | 60 | ||
| AK012369.1 | - | 5 - 6 | 487 | 60 | ||
| AK146186.1 | - | 5 - 6 | 512 | 60 | ||
| AK161330.1 | - | 5 - 6 | 460 | 60 | ||
| BC003775.1 | - | 5 - 6 | 511 | 3485097 | 60 | |
| HE864436.1 | - | 4 - 5 | 379 | 60 | ||
| X52959.1 | - | 5 - 6 | 492 | 60 | ||
| X56502.1 | - | 5 - 6 | 486 | 60 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Csnk2b
Genomic Map