Assay Details
| Gene Symbol: Csnk2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK002903.1 | - | 6 - 7 | 680 | 115 | |
| AK010730.1 | - | 6 - 7 | 677 | 115 | ||
| AK012369.1 | - | 6 - 7 | 675 | 115 | ||
| AK146186.1 | - | 6 - 7 | 700 | 115 | ||
| AK161330.1 | - | 6 - 7 | 648 | 115 | ||
| BC003775.1 | - | 6 - 7 | 699 | 3485097 | 115 | |
| HE864436.1 | - | 5 - 6 | 567 | 115 | ||
| HE864437.1 | - | 4 - 5 | 677 | 115 | ||
| X52959.1 | - | 6 - 7 | 680 | 115 | ||
| X56502.1 | - | 6 - 7 | 674 | 115 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Panther Classification:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31157712 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31157715 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31157717 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31157713 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31157720 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31157721 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31157714 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31157711 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31157710 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31157716 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P373379 | BC003775.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P373379 | X52959.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P373379 | X56502.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P373379 | AK010730.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P373379 | NM_009975.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P373379 | BC003775.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P373379 | X52959.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P373379 | X56502.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P373379 | AK010730.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P373379 | NM_009975.2 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 708669 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 971974 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 323238 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 35299 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P135740 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P373379 | BC003775.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P373379 | X52959.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P373379 | X56502.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P373379 | AK010730.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P373379 | NM_009975.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P373379 | BC003775.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P373379 | X52959.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P373379 | X56502.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P373379 | AK010730.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P373379 | NM_009975.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P373379 | BC003775.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P373379 | X52959.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P373379 | X56502.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P373379 | AK010730.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P373379 | NM_009975.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P373379 | BC003775.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P373379 | X52959.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P373379 | X56502.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P373379 | AK010730.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P373379 | NM_009975.2 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31157699 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31157729 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31157677 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31157707 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31157687 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31157708 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31157676 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31157701 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31157710 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31157689 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31157679 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31157720 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31157696 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31157680 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31157713 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31157715 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31157695 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31157686 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31157692 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31157705 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31157690 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31157725 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31157682 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31157722 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31157684 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31157685 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31157721 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31157717 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P135740 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08767528 | - | - |
| Whole Mouse Genome G4122A | A_51_P373379 | BC003775.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P373379 | X52959.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P373379 | X56502.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P373379 | AK010730.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P373379 | NM_009975.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P373379 | BC003775.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P373379 | X52959.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P373379 | X56502.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P373379 | AK010730.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P373379 | NM_009975.2 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P132720 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P135740 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13247237 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13127096 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13127103 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13127090 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13127089 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13505596 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13127095 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13127082 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13505599 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13127085 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31157715 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31157711 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31157720 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31157716 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31157717 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31157714 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31157712 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31157721 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31157713 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31157710 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31157677 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31157689 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31157783 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31157779 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31157639 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31157790 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31157674 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31157687 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31157670 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31157766 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31157675 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31157676 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31157666 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31157757 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31157729 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31157696 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31157668 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31157701 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31157679 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31157762 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31157722 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31157713 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31157615 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31157641 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31157680 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31157725 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31157637 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31157720 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31157708 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31157626 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31157684 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31157772 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31157659 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31157649 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31157747 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31157776 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31157690 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31157683 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31157787 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31157750 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31157634 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31157646 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31157692 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31157657 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31157769 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31157705 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31157764 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31157777 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31157655 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31157627 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31157621 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31157745 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31157717 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31157629 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31157609 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31157632 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31157752 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31157754 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31157654 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31157644 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31157770 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31157667 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31157780 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31157781 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31157700 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31157715 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31157625 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31157628 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31157710 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31157648 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31157613 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31157707 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31157611 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31157623 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31157789 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31157672 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31157679 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31157699 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31157676 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31157701 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31157700 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31157713 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13505596 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31157692 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13127101 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31157705 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31157717 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31157687 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13489064 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13127082 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31157682 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31157680 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31157715 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31157719 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31157722 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31157690 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31157689 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13506150 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31157710 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31157696 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31157707 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13506142 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10450438 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10450437 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10450441 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10450436 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10450439 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10450440 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10450443 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10450442 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1416728_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10450435 | - | - |
| Mouse Genome 430 2.0 Array | 1416728_at | - | - |
| Mouse Genome 430 2.0 Array | 1416728_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 13001_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 13001_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1700018481.mm.1 | NM_009975.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700018481.mm.1 | TC1700001947.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1700018482.mm.1 | NM_009975.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700018482.mm.1 | TC1700001947.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1416728_PM_at | - | - |
| Mouse Expression Set 430 | 1416728_at | - | - |
| Murine Genome U74v2 Set | 96099_PM_at | - | - |
| Murine Genome U74v2 Set | 96099_at | - | - |
| Mouse Genome 430A 2.0 Array | 1416728_at | - | - |
| Mouse Genome 430A 2.0 Array | 1416728_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1416728_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1416728_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 849727 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 5080161 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0005080161 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1224988 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1224988 | - | - |
| mouse-6 v1.1 (Illumina) | scl49996.9.1_82-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 5080161 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl49996.9.1_82-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 5080161 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2761215 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2761215 | - | - |

Csnk2b
Genomic Map