Assay Details
| Gene Symbol: Cyp24a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009996.3 | NP_034126.1 | 8 - 9 | 1519 | 91 | |
| GenBank mRNA | AK143984.1 | - | 8 - 9 | 1527 | 91 | |
| AK159109.1 | - | 8 - 9 | 1527 | 91 | ||
| AK159481.1 | - | 8 - 9 | 1527 | 91 | ||
| AK159527.1 | - | 8 - 9 | 1527 | 91 | ||
| AK159852.1 | - | 8 - 9 | 1527 | 91 | ||
| AK159954.1 | - | 8 - 9 | 1527 | 91 | ||
| AK159976.1 | - | 8 - 9 | 1527 | 91 | ||
| AK164697.1 | - | 8 - 9 | 1527 | 91 | ||
| BC109139.1 | - | 8 - 9 | 1511 | 40039500 | 91 | |
| BC109140.1 | - | 8 - 9 | 1511 | 40039502 | 91 | |
| D49438.1 | - | 8 - 9 | 1457 | 91 | ||
| D89669.1 | - | 8 - 9 | 1519 | 91 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cyp24a1
Genomic Map