Assay Details
| Gene Symbol: Fkbp4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010219.3 | NP_034349.1 | 4 - 5 | 638 | 61 | |
| GenBank mRNA | AK051433.1 | - | 664 | 61 | ||
| AK083912.1 | - | 4 - 5 | 638 | 61 | ||
| AK089972.1 | - | 4 - 5 | 61 | |||
| AK159211.1 | - | 4 - 5 | 665 | 61 | ||
| AK160202.1 | - | 4 - 5 | 587 | 61 | ||
| BC003447.1 | - | 4 - 5 | 655 | 2651490 | 61 | |
| X70887.1 | - | 4 - 5 | 535 | 61 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Fkbp4
Genomic Map