Assay Details
| Gene Symbol: Fkbp5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010220.4 | NP_034350.1 | 2 - 3 | 330 | 144 | |
| GenBank mRNA | AK169795.1 | - | 2 - 3 | 271 | 144 | |
| BC015260.1 | - | 2 - 3 | 269 | 4237766 | 144 | |
| BY740772.1 | - | 2 - 3 | 330 | 144 | ||
| CA317455.1 | - | 2 - 3 | 293 | 144 | ||
| CT010372.1 | - | 1 - 2 | 111 | 144 | ||
| U16959.1 | - | 2 - 3 | 330 | 144 | ||
| U36220.1 | - | 2 - 3 | 258 | 144 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Fkbp5
Genomic Map