Assay Details
| Gene Symbol: Fkbp5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010220.4 | NP_034350.1 | 8 - 9 | 1055 | 91 | |
| GenBank mRNA | AK169795.1 | - | 8 - 9 | 996 | 91 | |
| BC015260.1 | - | 8 - 9 | 994 | 4237766 | 91 | |
| BU613862.1 | - | 6 - 7 | 722 | 91 | ||
| CF949740.1 | - | 1 - 2 | 84 | 91 | ||
| CT010372.1 | - | 7 - 8 | 836 | 91 | ||
| U16959.1 | - | 8 - 9 | 1055 | 91 | ||
| U36220.1 | - | 8 - 9 | 983 | 91 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Fkbp5
Genomic Map