Assay Details
| Gene Symbol: Mxd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010751.3 | NP_034881.2 | 1 - 2 | 230 | 65 | |
| GenBank mRNA | AK036504.1 | - | 1 - 2 | 189 | 65 | |
| AK151014.1 | - | 1 - 2 | 192 | 65 | ||
| AK151114.1 | - | 1 - 2 | 191 | 65 | ||
| AK151305.1 | - | 1 - 2 | 229 | 65 | ||
| AK151882.1 | - | 1 - 2 | 191 | 65 | ||
| AK153303.1 | - | 1 - 2 | 191 | 65 | ||
| BC034860.1 | - | 1 - 2 | 167 | 1448816 | 65 | |
| BY106585.1 | - | 1 - 2 | 234 | 65 | ||
| CT010233.1 | - | 1 - 2 | 79 | 65 | ||
| U20614.1 | - | 1 - 2 | 184 | 65 | ||
| X83106.1 | - | 1 - 2 | 128 | 65 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mxd1
Genomic Map