Assay Details
| Gene Symbol: Mad1l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010752.3 | NP_034882.2 | 13 - 14 | 1475 | 73 | |
| GenBank mRNA | AF083812.1 | - | 13 - 14 | 1482 | 73 | |
| AF083813.1 | - | 13 - 14 | 1482 | 73 | ||
| AK136981.1 | - | 13 - 14 | 1478 | 73 | ||
| AK169976.1 | - | 2 - 3 | 444 | 73 | ||
| BC010702.1 | - | 13 - 14 | 1473 | 3156639 | 73 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mad1l1
Genomic Map