Assay Details
| Gene Symbol: Mag | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010758.2 | NP_034888.1 | 7 - 8 | 1383 | 63 | |
| GenBank mRNA | BC101944.1 | - | 7 - 8 | 1356 | 40039200 | 63 |
| BC103521.1 | - | 7 - 8 | 1356 | 40039204 | 63 | |
| BC103522.1 | - | 7 - 8 | 1356 | 63 | ||
| BC103523.1 | - | 8 - 9 | 1230 | 63 | ||
| M31811.1 | - | 7 - 8 | 1383 | 63 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mag
Genomic Map