Assay Details
| Gene Symbol: Man2b1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010764.2 | NP_034894.2 | 22 - 23 | 2883 | 67 | |
| GenBank mRNA | AK004817.2 | - | 22 - 23 | 2840 | 67 | |
| AK035551.1 | - | 22 - 23 | 2837 | 67 | ||
| AK147928.1 | - | 22 - 23 | 2841 | 67 | ||
| AK171166.1 | - | 22 - 23 | 2841 | 67 | ||
| BC005430.1 | - | 22 - 23 | 2828 | 3488681 | 67 | |
| BC023332.1 | - | 13 - 14 | 1585 | 67 | ||
| BC035318.1 | - | 9 - 10 | 1098 | 67 | ||
| U29947.1 | - | 22 - 23 | 2826 | 67 | ||
| U87240.1 | - | 22 - 23 | 2817 | 67 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Man2b1
Genomic Map