Assay Details
| Gene Symbol: Melk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010790.2 | NP_034920.2 | 8 - 9 | 864 | 101 | |
| GenBank mRNA | AK010498.1 | - | 8 - 9 | 830 | 101 | |
| AK011932.1 | - | 8 - 9 | 864 | 101 | ||
| AK129076.1 | - | 8 - 9 | 817 | 101 | ||
| AK137000.1 | - | 8 - 9 | 890 | 101 | ||
| AK145021.1 | - | 8 - 9 | 799 | 101 | ||
| AK145316.1 | - | 8 - 9 | 773 | 101 | ||
| AK161029.1 | - | 8 - 9 | 873 | 101 | ||
| AK164138.1 | - | 8 - 9 | 831 | 101 | ||
| AK167457.1 | - | 8 - 9 | 788 | 101 | ||
| BC020416.1 | - | 7 - 8 | 699 | 101 | ||
| BC085276.1 | - | 8 - 9 | 897 | 30443236 | 101 | |
| L76158.1 | - | 8 - 9 | 795 | 101 | ||
| X95351.1 | - | 8 - 9 | 802 | 101 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Melk
Genomic Map