Assay Details
| Gene Symbol: Minpp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010799.2 | NP_034929.1 | 3 - 4 | 1082 | 74 | |
| GenBank mRNA | AB041574.1 | - | 3 - 4 | 915 | 74 | |
| AF046908.1 | - | 3 - 4 | 1082 | 74 | ||
| AK128911.1 | - | 3 - 4 | 961 | 74 | ||
| AK151486.1 | - | 3 - 4 | 927 | 74 | ||
| BC021437.1 | - | 3 - 4 | 983 | 5039704 | 74 | |
More Information
Updated Mapping Information
Set Membership: |
|

Minpp1
Genomic Map