Assay Details
| Gene Symbol: Mnt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010813.3 | NP_034943.3 | 3 - 4 | 1102 | 72 | |
| GenBank mRNA | AK139743.1 | - | 2 - 3 | 124 | 72 | |
| AK147933.1 | - | 3 - 4 | 1086 | 72 | ||
| BC054534.1 | - | 3 - 4 | 1028 | 5721249 | 72 | |
| U77356.1 | - | 3 - 4 | 1102 | 72 | ||
| Y07609.1 | - | 3 - 4 | 1048 | 72 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mnt
Genomic Map