Assay Details
| Gene Symbol: Msh3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010829.2 | NP_034959.2 | 4 - 5 | 765 | 115 | |
| GenBank mRNA | AK040616.1 | - | 3 - 4 | 550 | 115 | |
| AK078467.1 | - | 4 - 5 | 716 | 115 | ||
| AK083388.1 | - | 4 - 5 | 671 | 115 | ||
| AK133907.1 | - | 4 - 5 | 757 | 115 | ||
| AK161089.1 | - | 4 - 5 | 760 | 116 | ||
| M24919.1 | - | 4 - 5 | 1048 | 115 | ||
| M80360.1 | - | 4 - 5 | 762 | 115 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Msh3
Genomic Map