Assay Details
| Gene Symbol: Slc25a15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_181325.4 | NP_851842.1 | 3 - 4 | 548 | 63 | |
| GenBank mRNA | AF133914.1 | - | 3 - 4 | 396 | 63 | |
| AK052758.1 | - | 3 - 4 | 480 | 63 | ||
| AK142223.1 | - | 3 - 4 | 418 | 63 | ||
| AK145966.1 | - | 3 - 4 | 418 | 63 | ||
| AK154988.1 | - | 3 - 4 | 524 | 63 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Slc25a15
Genomic Map