Assay Details
| Gene Symbol: Slc25a15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_181325.4 | NP_851842.1 | 5 - 6 | 863 | 64 | |
| GenBank mRNA | AF133914.1 | - | 5 - 6 | 711 | 64 | |
| AK052758.1 | - | 5 - 6 | 795 | 64 | ||
| AK081243.1 | - | 4 - 5 | 2987 | 64 | ||
| AK142223.1 | - | 5 - 6 | 733 | 64 | ||
| AK145966.1 | - | 5 - 6 | 733 | 64 | ||
| AK154988.1 | - | 5 - 6 | 840 | 64 | ||
| BC061143.1 | - | 5 - 6 | 504 | 30276598 | 64 | |
More Information
Updated Mapping Information
Set Membership: |
|

Slc25a15
Genomic Map