Assay Details
| Gene Symbol: St14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011176.4 | NP_035306.2 | 17 - 18 | 2440 | 77 | |
| GenBank mRNA | AF042822.3 | - | 17 - 18 | 2329 | 77 | |
| AK052738.1 | - | 17 - 18 | 2440 | 77 | ||
| AK166745.1 | - | 17 - 18 | 2431 | 77 | ||
| BC005496.1 | - | 17 - 18 | 2439 | 3488059 | 77 | |
| BC008514.1 | - | 17 - 18 | 2439 | 77 | ||
More Information
Updated Mapping Information
Set Membership: |
|

St14
Genomic Map