Assay Details
| Gene Symbol: Rad51d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001277938.1 | NP_001264867.1 | 5 - 6 | 687 | 69 | |
| NM_001277939.1 | NP_001264868.1 | 5 - 6 | 687 | 69 | ||
| NM_001277942.1 | NP_001264871.1 | 5 - 6 | 687 | 69 | ||
| NM_011235.4 | NP_035365.1 | 5 - 6 | 687 | 69 | ||
| NR_102717.1 | 5 - 6 | 687 | 69 | |||
| NR_102718.1 | 5 - 6 | 710 | 69 | |||
| NR_102719.1 | 4 - 5 | 568 | 69 | |||
| NR_102720.1 | 4 - 5 | 568 | 69 | |||
| GenBank mRNA | AB007040.1 | - | 5 - 6 | 652 | 69 | |
| AB052828.1 | - | 5 - 6 | 652 | 69 | ||
| AB052829.1 | - | 5 - 6 | 652 | 69 | ||
| AB052830.1 | - | 5 - 6 | 652 | 69 | ||
| AB052831.1 | - | 4 - 5 | 533 | 69 | ||
| AB052832.1 | - | 4 - 5 | 533 | 69 | ||
| AB052834.1 | - | 5 - 6 | 675 | 69 | ||
| AF034955.1 | - | 5 - 6 | 677 | 69 | ||
| AK017835.1 | - | 5 - 6 | 670 | 69 | ||
| AK138553.1 | - | 5 - 6 | 679 | 69 | ||
| AK147759.1 | - | 5 - 6 | 662 | 69 | ||
| BC049136.1 | - | 5 - 6 | 652 | 6832014 | 69 | |
| CF746344.1 | - | 1 - 2 | 104 | 69 | ||
| EU627687.1 | - | 5 - 6 | 677 | 69 | ||
| EU627688.1 | - | 5 - 6 | 677 | 69 | ||
| Y15570.1 | - | 5 - 6 | 673 | 69 | ||
More Information
Set Membership: |
|

Rad51d
Genomic Map