Assay Details
| Gene Symbol: Rad9a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011237.2 | NP_035367.1 | 6 - 7 | 649 | 58 | |
| GenBank mRNA | AF045663.1 | - | 6 - 7 | 649 | 58 | |
| AK134293.1 | - | 6 - 7 | 662 | 58 | ||
| AK138973.1 | - | 6 - 7 | 654 | 58 | ||
| AK165833.1 | - | 6 - 7 | 685 | 58 | ||
| BC082556.1 | - | 6 - 7 | 588 | 30538342 | 58 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rad9a
Genomic Map