Assay Details
| Gene Symbol: Rfc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011258.2 | NP_035388.2 | 2 - 3 | 249 | 91 | |
| GenBank mRNA | AK132028.1 | - | 2 - 3 | 229 | 91 | |
| AK136937.1 | - | 2 - 3 | 228 | 91 | ||
| AK146164.1 | - | 2 - 3 | 244 | 91 | ||
| AK146314.1 | - | 2 - 3 | 244 | 91 | ||
| BC085173.1 | - | 2 - 3 | 226 | 30549691 | 91 | |
| BC085174.1 | - | 2 - 3 | 218 | 91 | ||
| BY714128.1 | - | 2 - 3 | 277 | 91 | ||
| U01222.1 | - | 2 - 3 | 217 | 91 | ||
| U07157.1 | - | 2 - 3 | 173 | 91 | ||
| U15037.1 | - | 2 - 3 | 250 | 91 | ||
| U36441.1 | - | 2 - 3 | 226 | 91 | ||
| X72711.1 | - | 2 - 3 | 215 | 91 | ||
More Information
Set Membership: |
|

Rfc1
Genomic Map