Assay Details
| Gene Symbol: Sdf4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK157860.1 | - | 1 - 2 | 46 | 62 | |
| AK163400.1 | - | 1 - 2 | 166 | 62 | ||
| AK167113.1 | - | 1 - 2 | 95 | 62 | ||
| BC005715.1 | - | 2 - 3 | 317 | 62 | ||
| BC068152.1 | - | 1 - 2 | 421 | 30534982 | 62 | |
| BY226842.1 | - | 2 - 3 | 289 | 62 | ||
| D50461.1 | - | 1 - 2 | 424 | 62 | ||
| DN176224.1 | - | 1 - 2 | 492 | 62 | ||
| U45977.1 | - | 1 - 2 | 329 | 62 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AK157860.1,AK163400.1,AK167113.1,BC068152.1,D50461.1,U45977.1 transcripts.
Set Membership: |
|

Sdf4
Genomic Map