Assay Details
| Gene Symbol: Sftpc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011359.2 | NP_035489.2 | 2 - 3 | 241 | 81 | |
| GenBank mRNA | BC061137.1 | - | 2 - 3 | 236 | 30300758 | 81 |
| BY773034.1 | - | 2 - 3 | 242 | 81 | ||
| CJ093163.1 | - | 2 - 3 | 237 | 81 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Sftpc
Genomic Map