Assay Details
| Gene Symbol: Slc12a7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011390.2 | NP_035520.1 | 7 - 8 | 988 | 103 | |
| GenBank mRNA | AF087436.1 | - | 7 - 8 | 990 | 103 | |
| AK052910.1 | - | 7 - 8 | 1039 | 103 | ||
| AK083598.1 | - | 4 - 5 | 3259 | 103 | ||
| AK131129.1 | - | 7 - 8 | 942 | 103 | ||
| AK143535.1 | - | 7 - 8 | 982 | 103 | ||
| AK149750.1 | - | 7 - 8 | 983 | 103 | ||
| AK166215.1 | - | 7 - 8 | 982 | 103 | ||
| AK169950.1 | - | 7 - 8 | 103 | |||
| AK171498.1 | - | 7 - 8 | 1025 | 103 | ||
| BC059242.1 | - | 7 - 8 | 980 | 103 | ||
| BC060133.1 | - | 7 - 8 | 980 | 103 | ||
| BC079677.1 | - | 7 - 8 | 946 | 103 | ||
| BC141107.1 | - | 7 - 8 | 948 | 9055719 | 103 | |
More Information
Updated Mapping Information
Set Membership: |
|

Slc12a7
Genomic Map