Assay Details
| Gene Symbol: Slc12a7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011390.2 | NP_035520.1 | 17 - 18 | 2311 | 81 | |
| GenBank mRNA | AF087436.1 | - | 17 - 18 | 2313 | 81 | |
| AK131129.1 | - | 17 - 18 | 2265 | 81 | ||
| AK143535.1 | - | 17 - 18 | 2305 | 81 | ||
| AK149750.1 | - | 17 - 18 | 2306 | 81 | ||
| AK166215.1 | - | 17 - 18 | 2305 | 81 | ||
| AK169950.1 | - | 17 - 18 | 2301 | 81 | ||
| AK171498.1 | - | 17 - 18 | 2348 | 81 | ||
| BC059242.1 | - | 17 - 18 | 2303 | 81 | ||
| BC060133.1 | - | 17 - 18 | 2303 | 81 | ||
| BC079677.1 | - | 14 - 15 | 4061 | 81 | ||
| BC141107.1 | - | 17 - 18 | 2271 | 9055719 | 81 | |
More Information
Updated Mapping Information
Set Membership: |
|

Slc12a7
Genomic Map