Assay Details
| Gene Symbol: Slc12a7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011390.2 | NP_035520.1 | 19 - 20 | 2684 | 79 | |
| GenBank mRNA | AF087436.1 | - | 19 - 20 | 2686 | 79 | |
| AK131129.1 | - | 19 - 20 | 2638 | 79 | ||
| AK143535.1 | - | 19 - 20 | 2678 | 79 | ||
| AK149750.1 | - | 19 - 20 | 2679 | 79 | ||
| AK164405.1 | - | 2 - 3 | 1178 | 79 | ||
| AK166215.1 | - | 19 - 20 | 2678 | 79 | ||
| AK169950.1 | - | 19 - 20 | 2674 | 79 | ||
| AK171498.1 | - | 19 - 20 | 2721 | 79 | ||
| BC059242.1 | - | 19 - 20 | 2676 | 79 | ||
| BC060133.1 | - | 19 - 20 | 2676 | 79 | ||
| BC141107.1 | - | 19 - 20 | 2644 | 9055719 | 79 | |
More Information
Set Membership: |
|

Slc12a7
Genomic Map