Assay Details
| Gene Symbol: Slc25a14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011398.3 | NP_035528.1 | 2 - 3 | 491 | 109 | |
| GenBank mRNA | AF076981.1 | - | 2 - 3 | 492 | 109 | |
| CX238926.1 | - | 2 - 3 | 256 | 109 | ||
| CX244278.1 | - | 2 - 3 | 490 | 109 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AF076981.1,NM_011398.3 transcripts.
Set Membership: |
|

Slc25a14
Genomic Map