Assay Details
| Gene Symbol: Siglec1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011426.3 | NP_035556.3 | 12 - 13 | 3910 | 54 | |
| GenBank mRNA | BC141335.1 | - | 12 - 13 | 3504 | 9055947 | 54 |
| BC141336.1 | - | 12 - 13 | 3500 | 9055948 | 54 | |
| Z36233.1 | - | 12 - 13 | 3905 | 54 | ||
| Z36293.1 | - | 12 - 13 | 3905 | 54 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Siglec1
Genomic Map