Assay Details
| Gene Symbol: Siglec1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011426.3 | NP_035556.3 | 14 - 15 | 4435 | 101 | |
| GenBank mRNA | BC141335.1 | - | 14 - 15 | 4029 | 9055947 | 101 |
| BC141336.1 | - | 14 - 15 | 4025 | 9055948 | 101 | |
| Z36233.1 | - | 14 - 15 | 4430 | 101 | ||
| Z36293.1 | - | 14 - 15 | 4430 | 101 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Siglec1
Genomic Map