Assay Details
| Gene Symbol: Sox13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011439.2 | NP_035569.2 | 2 - 3 | 534 | 149 | |
| GenBank mRNA | AB006329.1 | - | 1 - 2 | 190 | 149 | |
| AJ000740.1 | - | 1 - 2 | 144 | 149 | ||
| BC007130.1 | - | 2 - 3 | 356 | 3493465 | 149 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2094731 | BE984765.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2094731 | BC007130.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2094731 | NM_011439.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2094731 | AB006329.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 405692 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 471073 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 675212 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 473388 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 657494 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 732090 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 382314 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 786255 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 309554 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 244901 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 543053 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 23492 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 567406 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 184700 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 718483 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 293745 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 782455 | AF332087.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 782455 | AF332088.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 782455 | AK052246.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 782455 | AK053428.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 782455 | AK150014.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 782455 | AK150075.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 782455 | BC058718.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 782455 | L15436.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 782455 | NM_001110204.1 | refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 782455 | NM_001110205.1 | refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 782455 | NM_007394.3 | refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 665337 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 441452 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 302645 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 453605 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 641889 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 644185 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 457448 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 263214 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 142232 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 96956 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 156666 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 117497 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P128938 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P163137 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2094731 | BE984765.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2094731 | BC007130.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2094731 | NM_011439.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2094731 | AB006329.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P520917 | BE984765.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P520917 | AJ000740.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P520917 | BC007130.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P520917 | NM_011439.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P520917 | AB006329.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P520917 | BE984765.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P520917 | AJ000740.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P520917 | BC007130.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P520917 | NM_011439.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P520917 | AB006329.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P110650 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P116548 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01110248 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01110255 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01110246 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P00163107 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P00163110 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01110250 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10357803 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10357794 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10357807 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10357801 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10357797 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10357799 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10357798 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10357804 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10357796 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10357806 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10357800 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10357791 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10357792 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10357795 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10357802 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1450191_PM_a_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10357790 | - | - |
| Mouse Genome 430 2.0 Array | 1450191_PM_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1450191_a_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 20668_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 20668_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0100024833.mm.1 | NM_011439.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100024833.mm.1 | NM_011439.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100024833.mm.1 | TC0100003090.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1450191_a_at | - | - |
| Mouse Expression Set 430 | 1450191_PM_a_at | - | - |
| Murine Genome U74v2 Set | 93698_at | - | - |
| Murine Genome U74v2 Set | 93698_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1450191_PM_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1450191_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1450191_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1450191_PM_a_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 928161 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 5690129 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 5690129 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0005690129 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1244292 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1244292 | - | - |
| mouse-6 v1.1 (Illumina) | scl16289.14.1_134-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 5690129 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl16289.14.1_134-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 5690129 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1244292 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1244292 | - | - |

Sox13
Genomic Map