Assay Details
| Gene Symbol: Sox6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001277326.1 | NP_001264255.1 | 8 - 9 | 1090 | 62 | |
| NM_011445.4 | NP_035575.1 | 9 - 10 | 1143 | 62 | ||
| GenBank mRNA | AK132409.1 | - | 8 - 9 | 1056 | 62 | |
| U32614.1 | - | 9 - 10 | 1147 | 62 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Sox6
Genomic Map