Assay Details
| Gene Symbol: Serpinb6b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011454.1 | NP_035584.1 | 1 - 2 | 29 | 105 | |
| GenBank mRNA | AK078753.1 | - | 1 - 2 | 99 | 105 | |
| AK084785.1 | - | 1 - 2 | 37 | 105 | ||
| AK154048.1 | - | 1 - 2 | 43 | 105 | ||
| BC061050.1 | - | 1 - 2 | 36 | 30276988 | 105 | |
| U96707.2 | - | 1 - 2 | 29 | 105 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AK084785.1,AK154048.1,BC061050.1,NM_011454.1,U96707.2 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2013043 | U96707.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2013043 | AK171741.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2013043 | NM_011454.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2013043 | AK078753.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 703690 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 819312 | AK087961.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 819312 | AK133952.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 819312 | AK145084.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 819312 | BC006728.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 819312 | NM_010849.4 | refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 819312 | X01023.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 747066 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 106194 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 880145 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 767590 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 38703 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 609915 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 877418 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 303683 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 86468 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 107708 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P106868 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P127182 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2013043 | U96707.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2013043 | AK171741.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2013043 | NM_011454.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2013043 | AK078753.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28932157 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28932193 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28932198 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28932196 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28932166 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28932155 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28932164 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28932184 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28932179 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28932174 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28932177 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28932195 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28932190 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28932181 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28932183 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28932194 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28932167 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28932182 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28932176 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28932200 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28932199 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28932186 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28932188 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28932187 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28932175 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28932159 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28932191 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28932180 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28932169 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28932172 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28932162 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28932154 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P02660659 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P106868 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07830900 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P127182 | - | - |
| Whole Mouse Genome G4122A | A_51_P288687 | U96707.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P288687 | AK171741.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P288687 | NM_011454.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P288687 | AK078753.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P127568 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P114476 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P127182 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P106868 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10545221 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10749932 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10749929 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10545224 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10545222 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28932164 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28932287 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28932242 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28932140 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28932258 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28932238 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28932305 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28932174 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28932208 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28932309 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28932196 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28932190 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28932181 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28932248 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28932272 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28932180 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28932285 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28932199 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28932179 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28932225 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28932203 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28932300 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28932211 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28932270 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28932227 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28932201 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28932265 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28932128 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28932263 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28932136 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28932149 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28932138 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28932132 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28932188 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28932215 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28932220 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28932194 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28932218 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28932312 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28932307 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28932273 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28932221 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28932253 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28932137 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28932296 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28932230 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28932250 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28932268 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28932167 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28932191 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28932286 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28932223 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28932130 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28932133 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28932217 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28932207 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28932157 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28932247 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28932124 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28932245 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28932175 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28932195 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28932292 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28932282 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28932166 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28932306 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28932159 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28932243 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28932241 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28932261 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28932303 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28932126 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28932276 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28932143 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28932232 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28932213 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28932289 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28932205 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28932154 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28932291 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28932155 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28932186 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28932297 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28932176 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28932184 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28932229 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28932224 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28932216 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28932262 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28932294 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28932271 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28932183 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28932191 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28932163 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28932175 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10749933 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10938313 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11110002 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10938308 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28932155 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28932167 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10749932 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11109986 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28932186 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11109996 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28932174 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28932177 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11002454 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28932172 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28932198 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28932187 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28932221 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28932166 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28932176 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28932154 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28932171 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28932210 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28932196 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28932143 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28932229 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28932227 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10938312 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10536406 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28932205 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28932211 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10749934 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28932195 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28932181 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28932180 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28932152 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28932188 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28932203 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28932230 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28932220 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11110004 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28932213 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28932179 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28932142 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28932206 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28932183 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28932151 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11110005 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28932223 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28932199 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28932159 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10545228 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10767507 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10817591 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28932149 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28932157 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10404424 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10404426 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10404423 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10404425 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10404427 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10404428 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1422804_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10404422 | - | - |
| Mouse Genome 430 2.0 Array | 1422804_at | - | - |
| Mouse Genome 430 2.0 Array | 1422804_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 20708_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 20708_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1300002140.mm.1 | NM_011454.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300002140.mm.1 | NM_011454.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300002140.mm.1 | TC1300000362.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1300002139.mm.1 | NM_011454.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300002139.mm.1 | NM_011454.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300002139.mm.1 | TC1300000362.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1300002141.mm.1 | NM_011454.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300002141.mm.1 | NM_011454.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300002141.mm.1 | TC1300000362.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1422804_at | - | - |
| Mouse Expression Set 430 | 1422804_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1422804_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1422804_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1422804_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1422804_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 609044 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 5700050 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 5700050 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0005700050 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2611755 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2611755 | - | - |
| mouse-6 v1.1 (Illumina) | scl44935.7_355-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 5700050 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl44935.7_355-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 5700050 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2611755 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2611755 | - | - |

Serpinb6b
Genomic Map