Assay Details
| Gene Symbol: Stk11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AB015801.1 | - | 4 - 5 | 635 | 71 | |
| AF129870.1 | - | 4 - 5 | 1013 | 71 | ||
| AF145287.1 | - | 4 - 5 | 617 | 71 | ||
| AF151711.1 | - | 4 - 5 | 617 | 71 | ||
| AK004499.1 | - | 2 - 3 | 296 | 71 | ||
| AK147802.1 | - | 4 - 5 | 676 | 71 | ||
| AK171909.1 | - | 5 - 6 | 1411 | 71 | ||
| AK172385.1 | - | 4 - 5 | 1400 | 71 | ||
| AK172528.1 | - | 4 - 5 | 1397 | 71 | ||
| BC052379.1 | - | 4 - 5 | 678 | 5688695 | 71 | |
| EU730638.1 | - | 4 - 5 | 596 | 71 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Stk11
Genomic Map