Assay Details
| Gene Symbol: Stk11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AB015801.1 | - | 5 - 6 | 768 | 67 | |
| AF129870.1 | - | 5 - 6 | 1146 | 67 | ||
| AF145287.1 | - | 5 - 6 | 750 | 67 | ||
| AF151711.1 | - | 5 - 6 | 750 | 67 | ||
| AK004499.1 | - | 3 - 4 | 429 | 67 | ||
| AK147802.1 | - | 5 - 6 | 810 | 67 | ||
| AK171909.1 | - | 6 - 7 | 1544 | 67 | ||
| AK172385.1 | - | 5 - 6 | 1533 | 67 | ||
| AK172528.1 | - | 5 - 6 | 1530 | 67 | ||
| BC052379.1 | - | 5 - 6 | 811 | 5688695 | 67 | |
| EU730638.1 | - | 5 - 6 | 729 | 67 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Stk11
Genomic Map