Assay Details
| Gene Symbol: Grem1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011824.4 | NP_035954.1 | 2 | 435 | 69 | |
| GenBank mRNA | AF045801.1 | - | 1 | 276 | 69 | |
| AF108189.1 | - | 1 | 345 | 69 | ||
| AK135022.1 | - | 2 | 470 | 69 | ||
| AK157793.1 | - | 2 | 436 | 69 | ||
| AK164871.1 | - | 2 | 436 | 69 | ||
| BC015293.1 | - | 2 | 405 | 4216669 | 69 | |
More Information
Updated Mapping Information
Set Membership: |
|

Grem1
Genomic Map