Assay Details
| Gene Symbol: Map2k5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011840.2 | NP_035970.1 | 1 - 2 | 728 | 62 | |
| GenBank mRNA | AB019374.1 | - | 1 - 2 | 729 | 62 | |
| AK020716.1 | - | 1 - 2 | 756 | 62 | ||
| AK159261.1 | - | 1 - 2 | 779 | 62 | ||
| AK166137.1 | - | 1 - 2 | 779 | 62 | ||
| BC013697.1 | - | 1 - 2 | 719 | 62 | ||
| BC028260.1 | - | 1 - 2 | 608 | 5067818 | 62 | |
More Information
Updated Mapping Information
Set Membership: |
|

Map2k5
Genomic Map