Assay Details
| Gene Symbol: Mta2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011842.3 | NP_035972.3 | 6 - 7 | 921 | 76 | |
| GenBank mRNA | AF159259.1 | - | 6 - 7 | 796 | 76 | |
| AK077548.1 | - | 6 - 7 | 903 | 76 | ||
| AK088158.1 | - | 6 - 7 | 834 | 76 | ||
| AK147099.1 | - | 6 - 7 | 900 | 76 | ||
| AK149834.1 | - | 6 - 7 | 967 | 76 | ||
| AK157717.1 | - | 6 - 7 | 825 | 76 | ||
| AK159979.1 | - | 6 - 7 | 897 | 76 | ||
| AK185749.1 | - | 4 - 5 | 333 | 76 | ||
| AK206505.1 | - | 3 - 4 | 319 | 76 | ||
| BC079847.1 | - | 6 - 7 | 921 | 6419411 | 76 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mta2
Genomic Map