Assay Details
| Gene Symbol: Mta2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011842.3 | NP_035972.3 | 15 - 16 | 2007 | 73 | |
| GenBank mRNA | AF159259.1 | - | 15 - 16 | 1882 | 73 | |
| AK003701.1 | - | 3 - 4 | 441 | 73 | ||
| AK077548.1 | - | 15 - 16 | 1989 | 73 | ||
| AK088158.1 | - | 15 - 16 | 1920 | 73 | ||
| AK147099.1 | - | 15 - 16 | 1986 | 73 | ||
| AK157717.1 | - | 15 - 16 | 1911 | 73 | ||
| AK159979.1 | - | 15 - 16 | 1983 | 73 | ||
| AK186550.1 | - | 1 - 2 | 33 | 73 | ||
| AK198824.1 | - | 1 - 2 | 33 | 73 | ||
| AK211099.1 | - | 2 - 3 | 246 | 73 | ||
| AK217458.1 | - | 1 - 2 | 97 | 73 | ||
| BC079847.1 | - | 15 - 16 | 2007 | 6419411 | 73 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mta2
Genomic Map