Assay Details
| Gene Symbol: Sept3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011889.2 | NP_036019.2 | 7 - 8 | 877 | 91 | |
| GenBank mRNA | AF104411.1 | - | 7 - 8 | 900 | 91 | |
| AK030668.1 | - | 7 - 8 | 845 | 91 | ||
| AK046722.1 | - | 7 - 8 | 970 | 91 | ||
| AK141915.1 | - | 7 - 8 | 804 | 91 | ||
| AK164861.1 | - | 7 - 8 | 974 | 91 | ||
| AK169916.1 | - | 7 - 8 | 998 | 91 | ||
| AK220520.1 | - | 5 - 6 | 676 | 91 | ||
| BC055738.1 | - | 7 - 8 | 806 | 6413697 | 91 | |
More Information
Updated Mapping Information
Set Membership: |
|

Sept3
Genomic Map