Assay Details
| Gene Symbol: Sgcg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011892.3 | NP_036022.1 | 6 - 7 | 730 | 82 | |
| GenBank mRNA | AB024922.1 | - | 7 - 8 | 835 | 82 | |
| AF282901.1 | - | 6 - 7 | 730 | 82 | ||
| AK132610.1 | - | 6 - 7 | 675 | 82 | ||
| BC138960.1 | - | 6 - 7 | 622 | 8861982 | 82 | |
| BC138961.1 | - | 6 - 7 | 622 | 8861983 | 82 | |
More Information
Set Membership: |
|

Sgcg
Genomic Map