Assay Details
| Gene Symbol: Slc9a3r1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_012030.2 | NP_036160.1 | 3 - 4 | 925 | 112 | |
| GenBank mRNA | AK085210.1 | - | 3 - 4 | 927 | 112 | |
| AK153558.1 | - | 3 - 4 | 925 | 112 | ||
| AK168894.1 | - | 3 - 4 | 926 | 112 | ||
| AK177064.1 | - | 1 - 2 | 105 | 112 | ||
| AK205456.1 | - | 1 - 2 | 105 | 112 | ||
| BC085141.1 | - | 3 - 4 | 910 | 6413140 | 112 | |
More Information
Updated Mapping Information
Set Membership: |
|

Slc9a3r1
Genomic Map