Assay Details
| Gene Symbol: Spo11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001083959.1 | NP_001077428.1 | 5 - 6 | 572 | 84 | |
| NM_001083960.1 | NP_001077429.1 | 5 - 6 | 533 | 84 | ||
| NM_012046.2 | NP_036176.1 | 6 - 7 | 647 | 84 | ||
| GenBank mRNA | AF126400.1 | - | 5 - 6 | 572 | 84 | |
| AF149309.1 | - | 5 - 6 | 496 | 84 | ||
| AF163053.1 | - | 5 - 6 | 533 | 84 | ||
| AF165313.1 | - | 5 - 6 | 526 | 84 | ||
| AF167439.1 | - | 5 - 6 | 565 | 84 | ||
| AF169386.1 | - | 6 - 7 | 644 | 84 | ||
| AF173848.1 | - | 6 - 7 | 640 | 84 | ||
| AK088498.1 | - | 3 - 4 | 1706 | 84 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Spo11
Genomic Map